Learn More
4-Oxocyclohexanecarboxylic Acid, TRC
Product Code. 30425592
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity Unit Size
30425592 1 g 1g
1 options
Product Code. 30425592 Supplier Toronto Research Chemicals Supplier No. TRCO8700751G
 Restricted Item
Forventet forsendelse: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Oxocyclohexanecarboxylic Acid
CAS 874-61-3
Molecular Formula C7H10O3
Quantity 1 g
Synonym 4-Ketocyclohexanecarboxylic Acid, 4-Ketocyclohexylcarboxylic Acid, 4-Oxo-1-cyclohexanecarboxylic Acid, 4-Oxocyclohexanecarboxylic Acid, Cyclohexanone-4-carboxylic Acid
InChI Formula InChI=1S/C7H10O3/c8-6-3-1-5(2-4-6)7(9)10/h5H,1-4H2,(H,9,10)
SMILES OC(=O)C1CCC(=O)CC1
IUPAC Name 4-oxocyclohexane-1-carboxylic acid
Molecular Weight (g/mol) 142.15
Formula Weight 142.063