Learn More
4-Oxo-4-(((R)-2,5,7,8-tetramethyl-2-((4R,8R)-4,8,12-trimethyltridecyl)chroman-6-yl)oxy)butanoic acid, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 4345-03-3 |
| Molecular Formula | C33H54O5 |
| MDL Number | MFCD00072055 |
| Quantity | 100 g |
| SMILES | O=C(O)CCC(OC1=C(C)C(CC[C@](CCC[C@H](C)CCC[C@H](C)CCCC(C)C)(C)O2)=C2C(C)=C1C)=O |
| Molecular Weight (g/mol) | 530.78 |
| Percent Purity | 98% |
| Chemical Name or Material | 4-Oxo-4-(((R)-2,5,7,8-tetramethyl-2-((4R,8R)-4,8,12-trimethyltridecyl)chroman-6-yl)oxy)butanoic acid |