Få mere at vide
4-Oxo-2,2,6,6-tetramethylpiperidine-d17, TRC
Artikelnummer. 30449384
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30449384 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30449384 Leverandør Toronto Research Chemicals Leverandørnr. TRCT3046315MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Oxo-2,2,6,6-tetramethylpiperidine-d17
CAS 52168-48-6
Molekylær formel C9 D17 N O
Mængde 5 mg
Synonym 4-Piperidinone-1,3,3,5,5-d5','2,2,6,6-tetra(methyl-d3)- (9CI','ACI), 2,2,6,6-Tetra(methyl-d3)-4-piperidinone-1,3,3,5,5-d5 (ACI), 2,2,6,6-Tetramethyl-4-oxo-1-piperidinyloxy-d16, Perdeuterated 2,2,6,6-tetramethyl-4-piperidone, 4-Oxo-2,2,6,6-tetramethylpiperidine-d17, 2,2,6,6-Tetramethyl-4-piperidone-D17
InChI formel InChI=1S/C9H17NO/c1-8(2)5-7(11)6-9(3,4)10-8/h10H,5-6H2,1-4H3/i1D3,2D3,3D3,4D3,5D2,6D2/hD
SMIL [2H]N1C(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])C(=O)C([2H])([2H])C1(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 1,3,3,5,5-pentadeuterio-2,2,6,6-tetrakis(trideuteriomethyl)piperidin-4-one
Molekylvægt (g/mol) 172.3421
Formel vægt 172.2377