Få mere at vide
4-Oxo-2,2,6,6-tetramethylpiperidine-d16-1-oxyl, CDN
Artikelnummer. 30319323
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30319323 0.25 g -
30315394 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319323 Leverandør CDN Leverandørnr. CDND23280.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,2,6,6-Tetramethyl-4-oxopiperidine-d16-1-oxyl
CAS 36763-53-8
Molekylær formel C9 2H16 N O2
Mængde 0.25 g
Synonym 1-Piperidinyl-3,3,5,5-d4-oxy, 2,2,6,6-tetra(methyl-d3)-4-oxo-, 2,2,6,6-Tetra(methyl-d3)-4-oxo-1-piperidinyl-3,3,5,5-d4-oxy, PD-Tempone, Perdeuterated 2,2,6,6-tetramethylpiperidone-N-oxyl, Perdeutero-2,2,6,6-tetramethyl-4-piperidone N-oxide, TANO-d16, 4-Oxo-TEMPO-d16, 4-Oxo-2,2,6,6-tetramethylpiperidine-d16-1-oxyl, 2,2,6,6-Tetramethyl-4-oxopiperidine-d16-1-oxyl, Tempone-d16, 1-λ1-Oxidanyl-2,2,6,6-tetramethylpiperidin-4-one-d16
InChI formel InChI=1 S/C9H16NO2/c1-8(2)5-7(11)6-9(3,4)10(8)12/h5-6H2,1-4H3/i1D3,2D3,3D3,4D3,5D2,6D2
SMIL [2 H]C([2 H])([2 H])C1(N([O])C(C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])C(=O)C1([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 3,3,5,5-tetradeuterio-1-λ^{1}-oxidanyl-2,2,6,6-tetrakis(trideuteriomethyl)piperidin-4-one
Molekylvægt (g/mol) 186.33
Formel vægt 186.2185 g/mol
Procent renhed 98 atom % D