Få mere at vide
4-Octylphenol-d4 Monoethoxylate (Major), TRC
Artikelnummer. 30417846
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30417846 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30417846 Leverandør Toronto Research Chemicals Leverandørnr. TRCO29377210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Octylphenol-d4 Monoethoxylate
CAS 1346602-86-5
Molekylær formel C16H22D4O2
Mængde 10 mg
Synonym 2-(4-Octylphenoxy)ethanol-d4, 2-(p-Octylphenoxy)ethanol-d4, Ethylene Glycol p-Octylphenyl-d4 Ether
InChI formel InChI=1S/C16H26O2/c1-2-3-4-5-6-7-8-15-9-11-16(12-10-15)18-14-13-17/h9-12,17H,2-8,13-14H2,1H3/i9D,10D,11D,12D
SMIL OCCOC1=C([2H])C([2H])=C(CCCCCCCC)C([2H])=C1[2H]
Molekylvægt (g/mol) 254.4
Formel vægt 254.2184