missing translation for 'onlineSavingsMsg'
Få mere at vide

STEMCELL Technologies 4-Octyl Itaconate

Artikelnummer. 17160342
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17160342 50 mg
17170342 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17160342 Leverandør STEMCELL Technologies Leverandørnr. 74192

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Activates nuclear factor erythroid 2-related factor 2 (Nrf2) protein

  • 4-Octyl Itaconate is a cell-permeable derivative of itaconate, a metabolite produced during the Krebs cycle
  • It is a potent activator of nuclear factor erythroid 2-related factor 2 (Nrf2) via alkylation of the inhibitor of Nrf2 (Keap1), thereby promoting Nrf2-mediated transcriptional activation and downstream anti-oxidant and anti-inflammatory properties (Mills et al.; Tang et al.)
  • The itaconate pathway (IRG1-itaconate-SDH) plays a pivotal role in up-regulating tolerance and inhibiting the induction of trained immunity (Domínguez-Andrés et al.)
  • Counteracts the proinflammatory signals of succinate by inhibiting succinate dehydrogenase (SDH) and activating Nrf2 to limit inflammation in mouse and human macrophages (Lampropoulou et al.; Mills et al.)
  • Activates Nrf2 signaling in human macrophages, which has been shown to inhibit the production of pro-inflammatory cytokines including TNF-α, IL-1β, and IL-6 (Tang et al.)

Tekniske data

CAS 3133-16-2
Synonym 2-methylene-butanedioic acid; 4-octyl ester
Mængde 50 mg
Molekylær formel C13H22O4
Renhed ≥98%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.