missing translation for 'onlineSavingsMsg'
Learn More

4-O-beta-Glucopyranosyl-cis-coumaric αcid, TRC

Product Code. 30401970
Change view
Click to view available options
Mængde:
2.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30401970 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30401970 Supplier Toronto Research Chemicals Supplier No. TRCG4190402.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 4-O-beta-Glucopyranosyl-cis-coumaric alphacid
CAS 117405-48-8
Molekylær formel C15H18O8
Mængde 2.5 mg
Synonym (2Z)-3-[4-(β-D-Glucopyranosyloxy)phenyl]-2-propenoic Acid, (Z)-p-Coumaric Acid 4-O-β-D-glucopyranoside, 4-O-β-D-Glucopyranosyl-cis-p-coumaric Acid, cis-p-Coumaric Acid 4-O-β-D-glucopyranoside
InChI formel InChI=1S/C15H18O8/c16-7-10-12(19)13(20)14(21)15(23-10)22-9-4-1-8(2-5-9)3-6-11(17)18/h1-6,10,12-16,19-21H,7H2,(H,17,18)/b6-3-/t10-,12-,13+,14-,15?/m1/s1
SMIL OC(/C=C\C1=CC=C(OC2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)C=C1)=O
IUPAC navn (Z)-3-[4-[(3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyphenyl]prop-2-enoic acid
Molekylvægt (g/mol) 326.3
Formel vægt 326.1002
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.