Få mere at vide
4-O-Acetyl-2,5-anhydro-1,3-O-isopropylidene-D-glucitol, TRC
Artikelnummer. 30324789
Skift visning
Mængde:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30324789 25 mg
30309963 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30324789 Leverandør Toronto Research Chemicals Leverandørnr. TRCA16837025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-O-Acetyl-2,5-anhydro-1,3-O-isopropylidene-D-glucitol
CAS 70128-28-8
Molekylær formel C11 H18 O6
Mængde 25 mg
InChI formel InChI=1S/C11H18O6/c1-6(13)15-9-7(4-12)16-8-5-14-11(2,3)17-10(8)9/h7-10,12H,4-5H2,1-3H3/t7-,8+,9-,10-/m1/s1
SMIL CC(=O)O[C@@H]1[C@@H](CO)O[C@H]2COC(C)(C)O[C@@H]12
IUPAC navn [(4aS,6R,7R,7aR)-6-(hydroxymethyl)-2,2-dimethyl-4a,6,7,7a-tetrahydro-4H-furo[3,2-d][1,3]dioxin-7-yl] acetate
Molekylvægt (g/mol) 246.26
Formel vægt 246.1103