missing translation for 'onlineSavingsMsg'
Få mere at vide

4-Nonyl Phenol-glucuronide, TRC

Artikelnummer. 30481000
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30481000 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30481000 Leverandør Toronto Research Chemicals Leverandørnr. TRCN65045550MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 4-Nonyl Phenol-glucuronide
CAS 207725-23-3
Molekylær formel C21 H32 O7
Mængde 50 mg
Synonym β-D-Glucopyranosiduronic acid 4-Nonylphenyl
InChI formel InChI=1S/C21H32O7/c1-2-3-4-5-6-7-8-9-14-10-12-15(13-11-14)27-21-18(24)16(22)17(23)19(28-21)20(25)26/h10-13,16-19,21-24H,2-9H2,1H3,(H,25,26)/t16-,17-,18+,19-,21+/m0/s1
SMIL CCCCCCCCCc1ccc(O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)O)cc1
IUPAC navn (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(4-nonylphenoxy)tetrahydropyran-2-carboxylic acid
Molekylvægt (g/mol) 396.47
Formel vægt 396.2148
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.