Få mere at vide
4-Nitropyridine-d4 N-Oxide, CDN
Artikelnummer. 30324254
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30324254 0.25 g -
30314890 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324254 Leverandør CDN Leverandørnr. CDND75970.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Nitropyridine N-Oxide-D4
CAS 119673-94-8
Molekylær formel C5 D4 N2 O3
Mængde 0.25 g
Synonym Pyridine-2,3,5,6-d4, 4-nitro-, 1-oxide (9 CI, ACI), 4-Nitropyridine-d4 N-Oxide, 4-Nitropyridine-2,3,5,6-d4 1-oxide, 4-Nitropyridine N-Oxide-D4
InChI formel InChI=1 S/C5H4N2O3/c8-6-3-1-5(2-4-6)7(9)10/h1-4 H/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])[n+]([O-])c([2 H])c([2 H])c1[N+](=O)[O-]
IUPAC navn 2,3,5,6-tetradeuterio-4-nitro-1-oxidopyridin-1-ium
Molekylvægt (g/mol) 144.1215
Formel vægt 144.0473 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity