missing translation for 'onlineSavingsMsg'
Få mere at vide

4-Nitrophenyl-Alpha-L-arabinofuranoside, TRC

Artikelnummer. 30480359
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30480359 10 mg
30481115 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30480359 Leverandør Toronto Research Chemicals Leverandørnr. TRCN50295010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 4-Nitrophenyl-alpha-L-arabinofuranoside
CAS 6892-58-6
Molekylær formel C11 H13 N O7
Mængde 10 mg
Synonym p-Nitrophenylarabinofuranoside, p-Nitrophenyl-α-L-arabinofuranoside
InChI formel InChI=1S/C11H13NO7/c13-5-8-9(14)10(15)11(19-8)18-7-3-1-6(2-4-7)12(16)17/h1-4,8-11,13-15H,5H2/t8-,9-,10+,11+/m0/s1
SMIL OC[C@@H]1O[C@@H](Oc2ccc(cc2)[N+](=O)[O-])[C@H](O)[C@H]1O
IUPAC navn (2S,3R,4R,5S)-2-(hydroxymethyl)-5-(4-nitrophenoxy)oxolane-3,4-diol
Molekylvægt (g/mol) 271.22
Formel vægt 271.0692
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.