Få mere at vide
4-Nitrophenyl 4,6-O-Benzylidene-Alpha-D-mannopyranoside, TRC
Artikelnummer. 30477474
Skift visning
Mængde:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30477474 50 mg
30477256 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30477474 Leverandør Toronto Research Chemicals Leverandørnr. TRCN49412550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Nitrophenyl 4,6-O-Benzylidene-a-D-mannopyranoside
CAS 58056-41-0
Molekylær formel C19 H19 N O8
Mængde 50 mg
InChI formel InChI=1S/C19H19NO8/c21-15-16(22)19(26-13-8-6-12(7-9-13)20(23)24)27-14-10-25-18(28-17(14)15)11-4-2-1-3-5-11/h1-9,14-19,21-22H,10H2/t14-,15-,16+,17-,18?,19+/m1/s1
SMIL O[C@@H]1[C@@H](Oc2ccc(cc2)[N+](=O)[O-])O[C@@H]3COC(O[C@H]3[C@@H]1O)c4ccccc4
IUPAC navn (4aR,6R,7S,8R,8aS)-6-(4-nitrophenoxy)-2-phenyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxine-7,8-diol
Molekylvægt (g/mol) 389.36
Formel vægt 389.1111