Få mere at vide
4-Nitrophenyl 3-O-Alpha-D-Glucopyranosyl-Alpha-D-glucopyranoside, TRC
Artikelnummer. 30479542
Skift visning
Mængde:
0.5 mg
1 mg
2 mg
Pakningsstørrelse:
0.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30479542 0.5 mg 0.5mg
30465500 1 mg -
30471807 2 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30479542 Leverandør Toronto Research Chemicals Leverandørnr. TRCN5036700.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 136632-95-6
Molekylær formel C18 H25 N O13
Molekylvægt (g/mol) 463.39
Synonym α-D-Glucopyranoside, 4-nitrophenyl 3-O-α-D-glucopyranosyl-, 4-Nitrophenyl 3-O-α-D-glucopyranosyl-α-D-glucopyranoside, 4-Nitrophenyl α-nigeroside, p-Nitrophenyl α-D-nigeroside
IUPAC navn (2R,3R,4S,5S,6R)-2-[(2R,3R,4S,5R,6R)-3,5-dihydroxy-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxan-4-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMIL OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](CO)O[C@H](Oc3ccc(cc3)[N+](=O)[O-])[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
Tekniske data
Kemisk navn eller materiale 4-Nitrophenyl 3-O-a-D-Glucopyranosyl-a-D-glucopyranoside
Mængde 0.5 mg
InChI formel InChI=1S/C18H25NO13/c20-5-9-11(22)13(24)14(25)17(30-9)32-16-12(23)10(6-21)31-18(15(16)26)29-8-3-1-7(2-4-8)19(27)28/h1-4,9-18,20-26H,5-6H2/t9-,10-,11-,12-,13+,14-,15-,16+,17-,18+/m1/s1
Formel vægt 463.1326