Få mere at vide
4-Nitrophenyl (2S)-2-(tert-Butoxycarbonylamino)-3-phenylpropanoate (N-BOC-L-Phenylalanine 4-Nitrophenyl Ester), Mikromol™
Beskrivelse
4-Nitrophenyl (2S)-2-(tert-Butoxycarbonylamino)-3-phenylpropanoate (N-BOC-L-Phenylalanine 4-Nitrophenyl Ester)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-Nitrophenyl (2S)-2-(tert-Butoxycarbonylamino)-3-phenylpropanoate |
| CAS | 7535-56-0 |
| Analyt- eller komponentnavne | 4-Nitrophenyl (2S)-2-(tert-Butoxycarbonylamino)-3-phenylpropanoate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C20 H22 N2 O6 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | L-Phenylalanine, N-[(1,1-dimethylethoxy)carbonyl]-, 4-nitrophenyl ester, Alanine, N-carboxy-3-phenyl-, N-tert-butyl p-nitrophenyl ester (7CI), Alanine, N-carboxy-3-phenyl-, N-tert-butyl p-nitrophenyl ester, L- (8CI), BOC-L-Phenylalanine p-nitrophenyl ester, N-(tert-Butoxycarbonyl)-L-phenylalanine p-nitrophenyl ester, N-(tert-Butoxycarbonyl)phenylalanine 4-nitrophenyl ester, N-(tert-Butoxycarbonyl)phenylalanine p-nitrophenyl ester, N-(tert-Butyloxycarbonyl)-L-phenylalanine 4-nitrophenyl ester, N-(tert-Butyloxycarbonyl)-L-phenylalanine p-nitrophenyl ester |
| InChI formel | InChI=1S/C20H22N2O6/c1-20(2,3)28-19(24)21-17(13-14-7-5-4-6-8-14)18(23)27-16-11-9-15(10-12-16)22(25)26/h4-12,17H,13H2,1-3H3,(H,21,24)/t17-/m0/s1 |
| SMIL | CC(C)(C)OC(=O)N[C@@H](Cc1ccccc1)C(=O)Oc2ccc(cc2)[N+](=O)[O-] |
| IUPAC navn | (4-nitrophenyl) (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoate |
| Vis mere |