missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl 2,3,4-tri-O-acetyl-Beta-D-glucuronic Acid, Methyl Ester, TRC

Product Code. 30478331
Change view
Click to view available options
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30478331 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478331 Supplier Toronto Research Chemicals Supplier No. TRCN506750250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 4-Nitrophenyl 2,3,4-tri-O-acetyl-beta-D-glucuronic Acid Methyl Ester
CAS 18472-49-6
Molekylær formel C19 H21 N O12
Mængde 250 mg
Synonym 4-Nitrophenyl 2,3,4-Tri-O-Acetyl-B-D-Glucuronic Acid, Methyl Ester
InChI formel InChI=1S/C19H21NO12/c1-9(21)28-14-15(29-10(2)22)17(30-11(3)23)19(32-16(14)18(24)27-4)31-13-7-5-12(6-8-13)20(25)26/h5-8,14-17,19H,1-4H3/t14-,15-,16-,17+,19+/m1/s1
SMIL COC(=O)[C@@H]1O[C@H](Oc2ccc(cc2)[N+](=O)[O-])[C@@H](OC(=O)C)[C@H](OC(=O)C)[C@H]1OC(=O)C
IUPAC navn methyl (2R,3R,4R,5S,6R)-3,4,5-triacetyloxy-6-(4-nitrophenoxy)oxane-2-carboxylate
Molekylvægt (g/mol) 455.37
Formel vægt 455.1064
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.