Få mere at vide
4-Nitrophenol-2,3,5,6-d4, CDN
Artikelnummer. 30321515
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30321515 0.25 g -
30320052 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321515 Leverandør CDN Leverandørnr. CDND23560.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Nitrophenol-2,3,5,6-D4
CAS 93951-79-2
Molekylær formel C6 2H4 H N O3
Mængde 0.25 g
Synonym 4-Nitrophenol-2,3,5,6-D4
InChI formel InChI=1 S/C6H5NO3/c8-6-3-1-5(2-4-6)7(9)10/h1-4,8 H/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])c(c([2 H])c([2 H])c1O)[N+](=O)[O-]
IUPAC navn 2,3,5,6-tetradeuterio-4-nitrophenol
Molekylvægt (g/mol) 143.13
Formel vægt 143.0521 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity