Få mere at vide
4-Nitrophenol-1,2,6-13C3, TRC
Artikelnummer. 30136676
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136676 50 mg
30009150 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30136676 Leverandør Toronto Research Chemicals Leverandørnr. TRCN49694850MG
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 93628-02-5
Molekylær formel 13C6H7NO3
Molekylvægt (g/mol) 147.08
Synonym Phenol-1,2,6-13C3, 4-nitro- (9CI), 4-Nitrophenol-1,2,6-13C3, p-Nitrophenol-13C3
IUPAC navn 4-(dihydroxyamino)(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-trien-1-ol
SMIL ON(O)[13c]1[13cH][13cH][13c](O)[13cH][13cH]1
Tekniske data
Kemisk navn eller materiale 4-Nitrophenol (13C6)
Mængde 50 mg
InChI formel InChI=1S/C6H7NO3/c8-6-3-1-5(2-4-6)7(9)10/h1-4,8-10H/i1+1,2+1,3+1,4+1,5+1,6+1
Formel vægt 147.0627