Få mere at vide
4-Nitrobutyric Acid Methyl Ester, TRC
Artikelnummer. 30479002
Skift visning
Mængde:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30479002 1 g
30471992 10 g
30465965 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30479002 Leverandør Toronto Research Chemicals Leverandørnr. TRCN4958101G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 13013-02-0
Molekylær formel C5H9NO4
Molekylvægt (g/mol) 147.13
Synonym 4-Nitrobutanoic Acid Methyl Ester, Methyl 4-Nitrobutanoate, Methyl 4-Nitrobutyrate, NSC 126459
IUPAC navn methyl 4-nitrobutanoate
SMIL COC(=O)CCC[N+](=O)[O-]
Tekniske data
Kemisk navn eller materiale 4-Nitrobutyric Acid Methyl Ester
Mængde 1 g
InChI formel InChI=1S/C5H9NO4/c1-10-5(7)3-2-4-6(8)9/h2-4H2,1H3
Formel vægt 147.0532