Få mere at vide
4-Nitrobiphenyl-d9, CDN
Artikelnummer. 30316919
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30316919 0.25 g
30316781 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316919 Leverandør CDN Leverandørnr. CDND50530.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Nitrobiphenyl-D9
CAS 350818-59-6
Molekylær formel C12 D9 N O2
Mængde 0.25 g
Synonym 1,1'-Biphenyl-2,2',3,3',4,5,5',6,6'-d9, 4'-nitro- (9 CI), 4'-Nitro-1,1'-biphenyl-2,2',3,3',4,5,5',6,6'-d9, 4-Nitrobiphenyl-d9, 4-Nitrobiphenyl-D9, 4-Nitrobiphenyl-2,2',3,3',4',5,5',6,6'-d9
InChI formel InChI=1 S/C12H9NO2/c14-13(15)12-8-6-11(7-9-12)10-4-2-1-3-5-10/h1-9 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D
SMIL [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])c2c([2 H])c([2 H])c(c([2 H])c2[2 H])[N+](=O)[O-]
IUPAC navn 1,2,3,4,5-pentadeuterio-6-(2,3,5,6-tetradeuterio-4-nitrophenyl)benzene
Molekylvægt (g/mol) 208.2608
Formel vægt 208.1198 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity