Få mere at vide
4-Nitrobenzonitrile-d4, CDN
Artikelnummer. 30321040
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30321040 0.1 g
30318070 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321040 Leverandør CDN Leverandørnr. CDND58960.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Nitrobenzonitrile-D4
CAS 1219798-46-5
Molekylær formel C7 D4 N2 O2
Mængde 0.1 g
Synonym Benzonitrile-2,3,5,6-d4, 4-nitro- (ACI), 4-Nitrobenzonitrile-2,3,5,6-d4 (ACI), 4-Nitrobenzonitrile-d4, 4-Nitrobenzonitrile-D4, p-Cyanonitrobenzene-d4, 1-Cyano-4-nitrobenzene-2,3,5,6-d4
InChI formel InChI=1 S/C7H4N2O2/c8-5-6-1-3-7(4-2-6)9(10)11/h1-4 H/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])c(c([2 H])c([2 H])c1C#N)[N+](=O)[O-]
IUPAC navn 2,3,5,6-tetradeuterio-4-nitrobenzonitrile
Molekylvægt (g/mol) 152.1435
Formel vægt 152.0524 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity