Få mere at vide
4-Nitrobenzoic Acid-d4, TRC
Artikelnummer. 30484053
Skift visning
Mængde:
25 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30484053 100 mg
30484166 25 mg
30464998 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30484053 Leverandør Toronto Research Chemicals Leverandørnr. TRCN493552100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 171777-66-5
Molekylær formel C7 D4 H N O4
Molekylvægt (g/mol) 171.14
Synonym p-Nitrobenzoic Acid-d4, NSC 7707-d4, Nitrodracylic Acid-d4, p-Carboxynitrobenzene-d4, p-Nitrobenzenecarboxylic Acid-d4, p-Nitrobenzoic Acid-d4, 4-Nitrobenzoic-d4 Acid
IUPAC navn 2,3,5,6-tetradeuterio-4-nitrobenzoic acid
SMIL [2H]c1c([2H])c(c([2H])c([2H])c1C(=O)O)[N+](=O)[O-]
Tekniske data
Kemisk navn eller materiale 4-Nitrobenzoic Acid-d4
Mængde 100 mg
InChI formel InChI=1S/C7H5NO4/c9-7(10)5-1-3-6(4-2-5)8(11)12/h1-4H,(H,9,10)/i1D,2D,3D,4D
Formel vægt 171.047