Få mere at vide
4-Nitroaniline-2,3,5,6-D4, TRC
Artikelnummer. 30473384
Skift visning
Mængde:
50 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30473384 50 mg
30470330 250 mg
30466133 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30473384 Leverandør Toronto Research Chemicals Leverandørnr. TRCN49177250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 64164-08-5
Molekylær formel C6 D4 H2 N2 O2
Molekylvægt (g/mol) 142.1487
Synonym Benzen-2,3,5,6-d4-amine, 4-nitro-, 4-Nitrobenzen-2,3,5,6-d4-amine, 2,3,5,6-Tetradeuterio-4-nitroaniline, 1-Amino-4-nitrobenzene-d4, 4-Nitroaniline-d4, p-Aminonitrobenzene-d4, p-Nitroaniline-d4
IUPAC navn 2,3,5,6-tetradeuterio-4-nitroaniline
SMIL [2H]c1c([2H])c(c([2H])c([2H])c1N)[N+](=O)[O-]
Tekniske data
Kemisk navn eller materiale 4-Nitroaniline-2,3,5,6-D4
Mængde 50 mg
InChI formel InChI=1S/C6H6N2O2/c7-5-1-3-6(4-2-5)8(9)10/h1-4H,7H2/i1D,2D,3D,4D
Formel vægt 142.068