Få mere at vide
4'-Nitroacetanilide-2',3',5',6'-d4, CDN
Artikelnummer. 30319730
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30319730 0.5 g 0.5g
30312881 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319730 Leverandør CDN Leverandørnr. CDND66210.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4'-Nitroacetanilide-2',3',5',6'-D4
CAS 68239-25-8
Molekylær formel C8 D4 H4 N2 O3
Mængde 0.5 g
Synonym Acetamide, N-(4-nitrophenyl-2,3,5,6-d4)- (9 CI, ACI), N-(4-Nitrophenyl-2,3,5,6-d4)acetamide (ACI), 4'-Nitroacetanilide-d4, 4'-Nitroacetanilide-2',3',5',6'-d4, 4'-Nitroacetanilide-2',3',5',6'-D4
InChI formel InChI=1 S/C8H8N2O3/c1-6(11)9-7-2-4-8(5-3-7)10(12)13/h2-5 H,1H3,(H,9,11)/i2D,3 D,4 D,5 D
SMIL [2 H]c1c([2 H])c(c([2 H])c([2 H])c1NC(=O)C)[N+](=O)[O-]
IUPAC navn N-(2,3,5,6-tetradeuterio-4-nitrophenyl)acetamide
Molekylvægt (g/mol) 184.1854
Formel vægt 184.0786 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity