Få mere at vide
4-Nitro-L-Phenylalanine Methyl Ester Hydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-Nitro-L-Phenylalanine Methyl Ester Hydrochloride |
| CAS | 17193-40-7 |
| Molekylær formel | C10H13ClN2O4 |
| Mængde | 10 g |
| Synonym | p-Nitrophenylalanine Methyl Ester Hydrochloride, (S)-1-Methoxycarbonyl-2-(4-nitrophenyl)ethanaminium Chloride, (S)-2-Amino-3-(4-nitrophenyl)propionic Acid Methyl Ester Hydrochloride, Methyl (S)-2-Amino-3-(4-nitrophenyl)propanoate Hydrochloride, Methyl 4-Nitro-L-phenylalaninate Hydrochloride |
| InChI formel | InChI=1S/C10H12N2O4.ClH/c1-16-10(13)9(11)6-7-2-4-8(5-3-7)12(14)15;/h2-5,9H,6,11H2,1H3;1H/t9-;/m0./s1 |
| SMIL | Cl.COC(=O)[C@@H](N)Cc1ccc(cc1)[N+](=O)[O-] |
| IUPAC navn | methyl (2S)-2-amino-3-(4-nitrophenyl)propanoate;hydrochloride |
| Molekylvægt (g/mol) | 260.67 |
| Formel vægt | 260.0564 |