Få mere at vide
4-Nitro-4’-aminodiphenyl-d4 Sulfone, TRC
Artikelnummer. 30473485
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30473485 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30473485 Leverandør Toronto Research Chemicals Leverandørnr. TRCN4915521MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Nitro-4'-aminodiphenyl-d4 Sulfone
CAS 1346601-85-1
Molekylær formel C12H6D4N2O4S
Mængde 1 mg
Synonym 4-(4-nitrobenzenesulfonyl)aniline-d4, 4-[(4-Nitrophenyl)sulfonyl]benzenamine-d4, 4-(4-Nitrobenzenesulfonyl)phenylamine-d4, 4-Nitro-4'-aminodiphenylsulfone-d4, NSC 27185-d5
InChI formel InChI=1S/C12H10N2O4S/c13-9-1-5-11(6-2-9)19(17,18)12-7-3-10(4-8-12)14(15)16/h1-8H,13H2/i1D,2D,5D,6D
SMIL [2H]c1c([2H])c(c([2H])c([2H])c1N)S(=O)(=O)c2ccc(cc2)[N+](=O)[O-]
IUPAC navn 2,3,5,6-tetradeuterio-4-(4-nitrophenyl)sulfonylaniline
Molekylvægt (g/mol) 282.31
Formel vægt 282.0612