Få mere at vide
4-Nitro-4’-acetylaminodiphenyl-d4 Sulfone, TRC
Artikelnummer. 30470322
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30470322 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30470322 Leverandør Toronto Research Chemicals Leverandørnr. TRCN49142710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Nitro-4'-acetylaminodiphenyl-d4 Sulfone
CAS 1346602-08-1
Molekylær formel C14H8D4N2O5S
Mængde 10 mg
Synonym N-[4-[(4-Nitrophenyl)sulfonyl]phenyl]acetamide-d4, 4'-[(p-Nitrophenyl)sulfonyl]acetanilide-d4, 4-Acetamido-4'-nitrodiphenyl-d4 Sulfone, NSC 30177-d4
InChI formel InChI=1S/C14H12N2O5S/c1-10(17)15-11-2-6-13(7-3-11)22(20,21)14-8-4-12(5-9-14)16(18)19/h2-9H,1H3,(H,15,17)/i2D,3D,6D,7D
SMIL [2H]c1c([2H])c(c([2H])c([2H])c1NC(=O)C)S(=O)(=O)c2ccc(cc2)[N+](=O)[O-]
IUPAC navn N-[2,3,5,6-tetradeuterio-4-(4-nitrophenyl)sulfonylphenyl]acetamide
Molekylvægt (g/mol) 324.35
Formel vægt 324.0718