missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-3-pyrazolecarboxylic acid, 95%, Thermo Scientific Chemicals

Catalog Number p-4089704
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5334-40-7
C4H3N3O4
157.08
MFCD00090899
ZMAXXOYJWZZQBK-UHFFFAOYSA-N
4-nitro-1h-pyrazole-3-carboxylic acid, 4-nitropyrazole-3-carboxylic acid, 4-nitro-3-pyrazolecarboxylic acid, 4-nitro-1h-pyrazole-5-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 4-nitro, 4-nitro-2h-pyrazole-3-carboxylic acid, 4-nitro pyrazole-3-carboxylic acid, 4-nitro-pyrazole-3-carboxylic acid, 4-hydroxynitroso-1h-pyrazole-3-carboxylic acid, nitropyrazolecarboxylic
219739
C1=NNC(=C1[N+](=O)[O-])C(=O)O
This item is not returnable. View return policy
5334-40-7
C4H3N3O4
157.08
MFCD00090899
ZMAXXOYJWZZQBK-UHFFFAOYSA-N
4-nitro-1h-pyrazole-3-carboxylic acid, 4-nitropyrazole-3-carboxylic acid, 4-nitro-3-pyrazolecarboxylic acid, 4-nitro-1h-pyrazole-5-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 4-nitro, 4-nitro-2h-pyrazole-3-carboxylic acid, 4-nitro pyrazole-3-carboxylic acid, 4-nitro-pyrazole-3-carboxylic acid, 4-hydroxynitroso-1h-pyrazole-3-carboxylic acid, nitropyrazolecarboxylic
219739
C1=NNC(=C1[N+](=O)[O-])C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 223.0°C to 225.0°C
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 157.08
Percent Purity 95%
Chemical Name or Material 4-Nitro-3-pyrazolecarboxylic acid
compliance-icons
Product Identifier
  • 4-Nitro-3-pyrazolecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'