missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Nitro-2-(trifluoromethoxy)aniline, 95%

Brand:  Alfa Aesar™ H63444.MD

Additional Details : CAS Number : 1261753-88-1 Weight : 0.00025kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Quantity 250mg

 View more versions of this product

Product Code. 15470139

  • 1207.92 DKK / 250mg


Estimated Shipment date
from Supplier 18-02-2019
Add to basket

Specifications

Chemical Identifiers

1261753-88-1
222.123
FFDBXRVBNGKCDQ-UHFFFAOYSA-N
56973585
C1=CC(=C(C=C1[N+](=O)[O-])OC(F)(F)F)N
C7H5F3N2O3
MFCD18399915
4-nitro-2-trifluoromethoxy aniline, 2-trifluoromethoxy-4-nitroaniline, 2-amino-5-nitrotrifluoromethoxybenzene
4-nitro-2-(trifluoromethoxy)aniline

Specifications

4-Nitro-2-(trifluoromethoxy)aniline
250mg
C7H5F3N2O3
UN2811
FFDBXRVBNGKCDQ-UHFFFAOYSA-N
4-nitro-2-(trifluoromethoxy)aniline
56973585
Powder
Yellow
1261753-88-1
MFCD18399915
4-nitro-2-trifluoromethoxy aniline, 2-trifluoromethoxy-4-nitroaniline, 2-amino-5-nitrotrifluoromethoxybenzene
C1=CC(=C(C=C1[N+](=O)[O-])OC(F)(F)F)N
222.123
222.12
95%
Videos
SDS