Få mere at vide
4-Nitro-2-phenoxyphenol, TRC
Artikelnummer. 30480595
Skift visning
Mængde:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30480595 25 mg
30466830 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30480595 Leverandør Toronto Research Chemicals Leverandørnr. TRCN49697825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 70995-08-3
Molekylær formel C12 H9 N O4
Molekylvægt (g/mol) 231.2
Synonym 4-Nitro-2-phenoxyphenol, Nimesulide Imp. G (EP)
IUPAC navn 4-nitro-2-phenoxyphenol
SMIL Oc1ccc(cc1Oc2ccccc2)[N+](=O)[O-]
Tekniske data
Kemisk navn eller materiale 4-Nitro-2-phenoxyphenol
Mængde 25 mg
InChI formel InChI=1S/C12H9NO4/c14-11-7-6-9(13(15)16)8-12(11)17-10-4-2-1-3-5-10/h1-8,14H
Formel vægt 231.0532