Få mere at vide
4-Nitro-2-phenoxyaniline, Mikromol™
Artikelnummer. 17982746
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17982746 25 mg
17950866 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17982746 Leverandør Mikromol Leverandørnr. MM0378.040025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-Nitro-2-phenoxyaniline
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Nitro-2-phenoxyaniline
CAS 5422-92-4
Analyt- eller komponentnavne 4-Nitro-2-phenoxyaniline
Forsendelsestilstand Room Temperature
Molekylær formel C12 H10 N2 O3
Certificeringer/compliance ISO 17025
Synonym 4-Nitro-2-phenoxyaniline, Nimesulide Imp. D (EP), Nimesulide Impurity D
InChI formel InChI=1S/C12H10N2O3/c13-11-7-6-9(14(15)16)8-12(11)17-10-4-2-1-3-5-10/h1-8H,13H2
SMIL Nc1ccc(cc1Oc2ccccc2)[N+](=O)[O-]
IUPAC navn 4-nitro-2-phenoxyaniline
Molekylvægt (g/mol) 230.22
Mængde 25 mg
Formel vægt 230.0691 g/mol