missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-n-Propylphenylacetylene, 97%

Brand:  Alfa Aesar™ B25065.22

Additional Details : CAS Number : 62452-73-7 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11496047

  • 7724.00 DKK / 100g

Estimated Shipment
28-05-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

62452-73-7
144.217
UVFFOABHOIMLNB-UHFFFAOYSA-N
2775133
CCCC1=CC=C(C=C1)C#C
C11H12
MFCD00173886
4-propylphenylacetylene, 1-eth-1-ynyl-4-propylbenzene, 4-n-propylphenylacetylene, 1-ethynyl-4-propyl-benzene, benzene, 1-ethynyl-4-propyl, pubchem10973, 4-n-propylphenyl ethine, 4-propyl phenylacetylene, 4-ethynyl-1-propylbenzene, 4-propyl-1-ethynylbenzene
1-ethynyl-4-propylbenzene

Specifications

4-n-Propylphenylacetylene
67°C (153°F)
205°C
62452-73-7
MFCD00173886
UVFFOABHOIMLNB-UHFFFAOYSA-N
1-ethynyl-4-propylbenzene
2775133
97%
0.91
1.534
100g
C11H12
4-propylphenylacetylene, 1-eth-1-ynyl-4-propylbenzene, 4-n-propylphenylacetylene, 1-ethynyl-4-propyl-benzene, benzene, 1-ethynyl-4-propyl, pubchem10973, 4-n-propylphenyl ethine, 4-propyl phenylacetylene, 4-ethynyl-1-propylbenzene, 4-propyl-1-ethynylbenzene
CCCC1=CC=C(C=C1)C#C
144.217
144.22
Videos
SDS Documents