Learn More
4-n-Propylacetophenone, TRC
Product Code. 30420674
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
Product Code. Quantity Unit Size
30420674 50 g 50g
1 options
Product Code. 30420674 Supplier Toronto Research Chemicals Supplier No. TRCP83225050G
 Restricted Item
Forventet forsendelse: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-n-Propylacetophenone
CAS 2932-65-2
Molecular Formula C11H14O
Quantity 50 g
Synonym 1-(4-Propylphenyl)ethanone, 1-Acetyl-4-propylbenzene, 4'-Propylacetophenone, Methyl 4-propylphenyl ketone, NSC 172873, p-Propylacetophenone, p-n-Propylacetophenone
InChI Formula InChI=1S/C11H14O/c1-3-4-10-5-7-11(8-6-10)9(2)12/h5-8H,3-4H2,1-2H3
SMILES CCCc1ccc(cc1)C(=O)C
IUPAC Name 1-(4-propylphenyl)ethanone
Molecular Weight (g/mol) 162.23
Formula Weight 162.1045