missing translation for 'onlineSavingsMsg'
Learn More

4-n-Propoxyphenyl trans-4-n-pentylcyclohexanecarboxylate, 98%, Thermo Scientific™

Catalog Number 15492726
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
67589-54-2
C21H32O3
332.484
MFCD09751071
ZAUPPIHFDVKVJM-UHFFFAOYSA-N
cyclohexanecarboxylic acid, 4-pentyl-, 4-propoxyphenyl ester, trans, 4-propoxyphenyl 4-pentylcyclohexane-1-carboxylate, trans-4-pentylcyclohexanecarboxylic acid, 4-propoxyphenyl ester, 4-propoxyphenyl 4-pentylcyclohexanecarboxylate, p-propoxyphenyl trans-4-pentylcyclohexanecarboxylate, 4-n-propoxyphenyl trans-4-n-pentylcyclohexanecarboxylate, 4-pentyl-1-cyclohexanecarboxylic acid 4-propoxyphenyl ester, trans-4-pentylcyclohexanecarboxylic acid 4-propoxyphenyl ester, 4-propoxyphenyl 1s,4r-4-pentylcyclohexane-1-carboxylate, 4alpha-pentyl-1beta-cyclohexanecarboxylic acid 4-propyloxy phenyl ester
105461
(4-propoxyphenyl) 4-pentylcyclohexane-1-carboxylate
CCCCCC1CCC(CC1)C(=O)OC2=CC=C(C=C2)OCCC
This item is not returnable. View return policy
67589-54-2
C21H32O3
332.484
MFCD09751071
ZAUPPIHFDVKVJM-UHFFFAOYSA-N
cyclohexanecarboxylic acid, 4-pentyl-, 4-propoxyphenyl ester, trans, 4-propoxyphenyl 4-pentylcyclohexane-1-carboxylate, trans-4-pentylcyclohexanecarboxylic acid, 4-propoxyphenyl ester, 4-propoxyphenyl 4-pentylcyclohexanecarboxylate, p-propoxyphenyl trans-4-pentylcyclohexanecarboxylate, 4-n-propoxyphenyl trans-4-n-pentylcyclohexanecarboxylate, 4-pentyl-1-cyclohexanecarboxylic acid 4-propoxyphenyl ester, trans-4-pentylcyclohexanecarboxylic acid 4-propoxyphenyl ester, 4-propoxyphenyl 1s,4r-4-pentylcyclohexane-1-carboxylate, 4alpha-pentyl-1beta-cyclohexanecarboxylic acid 4-propyloxy phenyl ester
105461
(4-propoxyphenyl) 4-pentylcyclohexane-1-carboxylate
CCCCCC1CCC(CC1)C(=O)OC2=CC=C(C=C2)OCCC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 332.48
Percent Purity 98%
Chemical Name or Material 4-n-Propoxyphenyl trans-4-n-pentylcyclohexanecarboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'