missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4'-n-Propoxybiphenyl-4-boronic acid, 97%

Brand:  Alfa Aesar™ H55999.03

Additional Details : CAS Number : 849062-20-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15482408

  • 4430.03 DKK / 1g


Estimated Shipment
26-10-2020
Add to basket

Specifications

Chemical Identifiers

849062-20-0
256.108
ZJCHLFOLMBDUPR-UHFFFAOYSA-N
16217483
B(C1=CC=C(C=C1)C2=CC=C(C=C2)OCCC)(O)O
C15H17BO3
MFCD06798061
4-4'-propoxyphenyl phenylboronic acid, 4'-propoxy-1,1'-biphenyl-4-yl boronic acid, 4-4-propoxyphenyl phenylboronic acid, 4'-propoxy-1,1'-biphenyl-4-ylboronic acid, 4'-propoxy-1,1'-biphenyl-4-yl boronicacid, 4'-propoxy 1,1'-biphenyl-4-yl boronic acid, 4-4 inverted exclamation marka-propoxyphenyl phenylboronic acid
[4-(4-propoxyphenyl)phenyl]boronic acid

Specifications

4'-n-Propoxybiphenyl-4-boronic acid
849062-20-0
MFCD06798061
ZJCHLFOLMBDUPR-UHFFFAOYSA-N
[4-(4-propoxyphenyl)phenyl]boronic acid
16217483
Solid
1g
C15H17BO3
4-4'-propoxyphenyl phenylboronic acid, 4'-propoxy-1,1'-biphenyl-4-yl boronic acid, 4-4-propoxyphenyl phenylboronic acid, 4'-propoxy-1,1'-biphenyl-4-ylboronic acid, 4'-propoxy-1,1'-biphenyl-4-yl boronicacid, 4'-propoxy 1,1'-biphenyl-4-yl boronic acid, 4-4 inverted exclamation marka-propoxyphenyl phenylboronic acid
B(C1=CC=C(C=C1)C2=CC=C(C=C2)OCCC)(O)O
256.108
256.11
97%
Videos
SDS Documents