Få mere at vide
4-n-Pentylphenol-2,3,5,6-d4,OD, CDN
Artikelnummer. 30319547
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30319547 0.05 g -
30314888 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319547 Leverandør CDN Leverandørnr. CDND58570.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Pentylphenol-O,2,3,5,6-D5
CAS 126839-95-0
Molekylær formel C11 D5 H11 O
Mængde 0.05 g
Synonym Phen-2,3,5,6-d4-ol-d, 4-pentyl- (9 CI), 4-Pentylphenol-O,2,3,5,6-D5, 4-n-Pentylphenol-2,3,5,6-d4,OD, 4-n-Amylphenol D5 (phenol D5), 4-Pentylphen-2,3,5,6-d4-ol-d
InChI formel InChI=1 S/C11H16O/c1-2-3-4-5-10-6-8-11(12)9-7-10/h6-9,12 H,2-5H2,1H3/i6D,7 D,8 D,9 D/hD
SMIL [2 H]Oc1c([2 H])c([2 H])c(CCCCC)c([2 H])c1[2 H]
IUPAC navn 1,2,4,5-tetradeuterio-3-deuteriooxy-6-pentylbenzene
Molekylvægt (g/mol) 169.275
Formel vægt 169.1515 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity