Få mere at vide
4-n-Octyl-d17-phenol, TRC
Artikelnummer. 30411878
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30411878 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30411878 Leverandør Toronto Research Chemicals Leverandørnr. TRCO2937761MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-n-Octyl-d17-phenol
CAS 1219794-55-4
Molekylær formel C14 D17 H5 O
Mængde 1 mg
Synonym 4-Octylphenol D17 (octyl D17)
InChI formel InChI=1S/C14H22O/c1-2-3-4-5-6-7-8-13-9-11-14(15)12-10-13/h9-12,15H,2-8H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])c1ccc(O)cc1
IUPAC navn 4-(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecadeuteriooctyl)phenol
Molekylvægt (g/mol) 223.43
Formel vægt 223.2738