Få mere at vide
(4-n-Nonylphenoxy)acetic-Alpha,Alpha-d2 Acid, TRC
Artikelnummer. 30475153
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30475153 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30475153 Leverandør Toronto Research Chemicals Leverandørnr. TRCN6504615MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Nonylphenoxy-aceticacidD2
CAS 1219798-75-0
Molekylær formel C17 2H2 H24 O3
Mængde 5 mg
Synonym 4-Nonylphenoxy Acetic Acid D2 (2-D2), 4-Nonylphenoxy-acetic acid D2
InChI formel InChI=1S/C17H26O3/c1-2-3-4-5-6-7-8-9-15-10-12-16(13-11-15)20-14-17(18)19/h10-13H,2-9,14H2,1H3,(H,18,19)/i14D2
SMIL [2H]C([2H])(Oc1ccc(CCCCCCCCC)cc1)C(=O)O
IUPAC navn 2,2-dideuterio-2-(4-nonylphenoxy)acetic acid
Molekylvægt (g/mol) 280.4
Formel vægt 280.2007