Få mere at vide
(4-n-Nonylphenoxy)acetic-alpha,alpha-d2 Acid, CDN
Artikelnummer. 30323923
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30323923 0.1 g
30318781 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323923 Leverandør CDN Leverandørnr. CDND64920.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Nonylphenoxy-aceticacidD2
CAS 1219798-75-0
Molekylær formel C17 2H2 H24 O3
Mængde 0.1 g
InChI formel InChI=1 S/C17H26O3/c1-2-3-4-5-6-7-8-9-15-10-12-16(13-11-15)20-14-17(18)19/h10-13 H,2-9,14H2,1H3,(H,18,19)/i14D2
SMIL [2 H]C([2 H])(Oc1ccc(CCCCCCCCC)cc1)C(=O)O
IUPAC navn 2,2-dideuterio-2-(4-nonylphenoxy)acetic acid
Molekylvægt (g/mol) 280.4
Formel vægt 280.2007 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity