Få mere at vide
4-n-Nonylphenol-2,3,5,6-d4, TRC
Artikelnummer. 30466164
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30466164 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30466164 Leverandør Toronto Research Chemicals Leverandørnr. TRCN65043425MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-n-Nonylphenol D4 (ring D4)
CAS 1173019-62-9
Renhedsgrad noter HPLC
Molekylær formel C15 2H4 H20 O
Mængde 25 mg
Synonym Phen-2,3,5,6-D4-ol','4-nonyl-, 4-n-Nonylphenol D4 (ring D4), 4-Nonylphen-2,3,4,5-d4-ol (ACI), Phen-2,3,5,6-D4-ol','4-nonyl-
InChI formel InChI=1S/C15H24O/c1-2-3-4-5-6-7-8-9-14-10-12-15(16)13-11-14/h10-13,16H,2-9H2,1H3/i10D,11D,12D,13D
SMIL [2H]c1c([2H])c(CCCCCCCCC)c([2H])c([2H])c1O
IUPAC navn 2,3,5,6-tetradeuterio-4-nonylphenol
Molekylvægt (g/mol) 224.38
Formel vægt 224.2078
Procent renhed >95