Få mere at vide
4-n-Nonylphenol-2,3,5,6-d4,OD, CDN
Artikelnummer. 30319880
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30319880 0.25 g -
30309747 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319880 Leverandør CDN Leverandørnr. CDND55100.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale p-n-Nonylphenol phenol-d5
CAS 358730-95-7
Molekylær formel C15 D5 H19 O
Mængde 0.25 g
Synonym Phen-2,3,5,6-d4-ol-d, 4-nonyl- (9 CI, ACI), 4-Nonylphen-2,3,5,6-d4-ol-d (ACI), 4-n-Nonylphenol-2,3,5,6-d4,OD, 4-n-Nonylphenol-O,2,3,5,6-D5
InChI formel InChI=1 S/C15H24O/c1-2-3-4-5-6-7-8-9-14-10-12-15(16)13-11-14/h10-13,16 H,2-9H2,1H3/i10D,11 D,12 D,13 D/hD
SMIL [2 H]Oc1c([2 H])c([2 H])c(CCCCCCCCC)c([2 H])c1[2 H]
IUPAC navn 1,2,4,5-tetradeuterio-3-deuteriooxy-6-nonylbenzene
Molekylvægt (g/mol) 225.3813
Formel vægt 225.2141 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity