missing translation for 'onlineSavingsMsg'
Få mere at vide

4-(N-Methyl-N-nitrosamino)-4-(3-pyridyl)butanal, TRC

Artikelnummer. 30136577
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2.5 mg
5 mg
10 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30136577 2.5 mg 2.5mg
18032250 10 mg -
18082840 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 30136577 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3256502.5MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64091-90-3
Molekylær formel C10 H13 N3 O2
Molekylvægt (g/mol) 207.23
IUPAC navn N-methyl-N-(4-oxo-1-pyridin-3-ylbutyl)nitrous amide
SMIL CN(N=O)C(CCC=O)c1cccnc1

Specifications

Kemisk navn eller materiale 4-(N-Methyl-N-nitrosamino)-4-(3-pyridyl)butanal
Mængde 2.5 mg
InChI formel InChI=1S/C10H13N3O2/c1-13(12-15)10(5-3-7-14)9-4-2-6-11-8-9/h2,4,6-8,10H,3,5H2,1H3
Formel vægt 207.1008
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.