Få mere at vide
4-[N-(Methyl-d3)-N-nitrosamino]-4-(3-pyridyl)butanal, TRC
Artikelnummer. 30464340
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30464340 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30464340 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3256521MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-[N-(Methyl-d3)-N-nitrosamino]-4-(3-pyridyl)butanal
CAS 1185039-70-6
Molekylær formel C10H10D3N3O2
Mængde 1 mg
Synonym γ-[(Methyl-d3)nitrosoamino]-3-pyridinebutanal, 4-[N-(Methyl-d3)-N-nitrosamino]-4-(3-pyridyl)-1-butanal, NNA-d3
InChI formel InChI=1S/C10H13N3O2/c1-13(12-15)10(5-3-7-14)9-4-2-6-11-8-9/h2,4,6-8,10H,3,5H2,1H3/i1D3
SMIL O=CCCC(N(N=O)C([2H])([2H])[2H])C1=CN=CC=C1
Molekylvægt (g/mol) 210.25
Formel vægt 210.1196