missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-n-Hexylbenzeneboronic acid, 97%

Brand:  Alfa Aesar™ H64555.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15471829

  • 1334.97 DKK / 1g


In stock, ready to ship: 1
Add to basket

Specifications

Chemical Identifiers

105365-50-2
206.092
CFYBKGAFWFRFRJ-UHFFFAOYSA-N
4197864
B(C1=CC=C(C=C1)CCCCCC)(O)O
C12H19BO2
MFCD04039029
4-hexylphenyl boronic acid, 4-n-hexylphenylboronic acid, 4-hex-1-yl benzeneboronic acid, 1-borono-4-hex-1-yl benzene, 4-n-hexylbenzeneboronic acid, boronic acid, 4-hexylphenyl, 4-hexylphenylboronicacid, p-hexylphenylboronic acid, 4-hexylbenzeneboronic acid, 4-hexylphenyl-boronic acid
(4-hexylphenyl)boronic acid

Specifications

4-n-Hexylbenzeneboronic acid
105365-50-2
MFCD04039029
CFYBKGAFWFRFRJ-UHFFFAOYSA-N
(4-hexylphenyl)boronic acid
4197864
Solid
1g
C12H19BO2
4-hexylphenyl boronic acid, 4-n-hexylphenylboronic acid, 4-hex-1-yl benzeneboronic acid, 1-borono-4-hex-1-yl benzene, 4-n-hexylbenzeneboronic acid, boronic acid, 4-hexylphenyl, 4-hexylphenylboronicacid, p-hexylphenylboronic acid, 4-hexylbenzeneboronic acid, 4-hexylphenyl-boronic acid
B(C1=CC=C(C=C1)CCCCCC)(O)O
206.092
206.09
97%
Videos
SDS Documents