Få mere at vide
4-n-Butylphenol-2,3,5,6-d4,OD, CDN
Artikelnummer. 30322111
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30322111 0.1 g 0.1g
30317490 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322111 Leverandør CDN Leverandørnr. CDND59300.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Butylphenol-O,2,3,5,6-D5
CAS 1219795-04-6
Molekylær formel C10 D5 H9 O
Mængde 0.1 g
Synonym 4-n-Butylphenol-2,3,5,6-d4,OD, 4-Butylphenol-O,2,3,5,6-D5, p-(Hydroxy-d)-n-butylbenzene-2,3,5,6-d4, 4-Butylphenol-d5
InChI formel InChI=1 S/C10H14O/c1-2-3-4-9-5-7-10(11)8-6-9/h5-8,11 H,2-4H2,1H3/i5D,6 D,7 D,8 D/hD
SMIL [2 H]Oc1c([2 H])c([2 H])c(CCCC)c([2 H])c1[2 H]
IUPAC navn 1-butyl-2,3,5,6-tetradeuterio-4-deuteriooxybenzene
Molekylvægt (g/mol) 155.2484
Formel vægt 155.1358 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity