missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butoxybenzaldehyde, 98%, Thermo Scientific™

Catalog Number p-7022455
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
5736-88-9
C11H14O2
178.231
MFCD00003389
XHWMNHADTZZHGI-UHFFFAOYSA-N
p-butoxybenzaldehyde, benzaldehyde, 4-butoxy, 4-n-butoxybenzaldehyde, benzaldehyde, p-butoxy, 4-butoxy-benzaldehyde, p-n-butoxybenzaldehyde, 4-n-butyloxybenzaldehyde, acmc-209lxn, p-n-butoxy benzaldehyde, 4-butoxybenzaldehyde
79813
4-butoxybenzaldehyde
CCCCOC1=CC=C(C=C1)C=O
This item is not returnable. View return policy
5736-88-9
C11H14O2
178.231
MFCD00003389
XHWMNHADTZZHGI-UHFFFAOYSA-N
p-butoxybenzaldehyde, benzaldehyde, 4-butoxy, 4-n-butoxybenzaldehyde, benzaldehyde, p-butoxy, 4-butoxy-benzaldehyde, p-n-butoxybenzaldehyde, 4-n-butyloxybenzaldehyde, acmc-209lxn, p-n-butoxy benzaldehyde, 4-butoxybenzaldehyde
79813
4-butoxybenzaldehyde
CCCCOC1=CC=C(C=C1)C=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 1.031
Boiling Point 284°C to 286°C
Flash Point >110°C (230°F)
Refractive Index 1.541
Quantity 10 g
Sensitivity Air Sensitive
Formula Weight 178.23
Percent Purity 98%
Chemical Name or Material 4-n-Butoxybenzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'