missing translation for 'onlineSavingsMsg'
Learn More

4-Morpholinophenyl isothiocyanate, Thermo Scientific™

Catalog Number p-4510163
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
51317-66-9
C11H12N2OS
220.29
AXUXRZZYZBZQAR-UHFFFAOYSA-N
224862
4-(4-isothiocyanatophenyl)morpholine
C1COCCN1C2=CC=C(C=C2)N=C=S
This item is not returnable. View return policy
51317-66-9
C11H12N2OS
220.29
AXUXRZZYZBZQAR-UHFFFAOYSA-N
224862
4-(4-isothiocyanatophenyl)morpholine
C1COCCN1C2=CC=C(C=C2)N=C=S
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 4-morpholinophenyl isothiocyanate, 4-4-isothiocyanatophenyl morpholine, 4-morpholinophenylisothiocyanate, morpholine, 4-4-isothiocyanatophenyl, 4-4-isothiocyanatophenyl morpholin, 4-morpholin-4-ylbenzenisothiocyanate, 4-4-morpholinyl phenyl isothiocyanate, 4-4-isothiocyanato-phenyl-morpholine, morpholine,4-4-isothiocyanatophenyl, 4-morpholin-4-yl phenyl isothiocyanate
Formula Weight 220.29
Percent Purity 97%
Chemical Name or Material 4-Morpholinophenyl isothiocyanate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'