missing translation for 'onlineSavingsMsg'
Learn More

4-Morpholinoaniline, 97%, Thermo Scientific™

Catalog Number p-2702034
Click to view available options
Quantity:
10 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
2524-67-6
C10H14N2O
178.235
MFCD00006169
PHNDZBFLOPIMSM-UHFFFAOYSA-N
4-morpholinoaniline, 4-morpholin-4-yl-phenylamine, 4-morpholin-4-yl aniline, n-4-aminophenyl morpholine, p-morpholinoaniline, 4-morpholinobenzenamine, 4-4-morpholino aniline, benzenamine, 4-4-morpholinyl, 4-morpholinyl aniline, 4-4-aminophenyl-morpholine
75655
4-morpholin-4-ylaniline
C1COCCN1C2=CC=C(C=C2)N
This item is not returnable. View return policy
2524-67-6
C10H14N2O
178.235
MFCD00006169
PHNDZBFLOPIMSM-UHFFFAOYSA-N
4-morpholinoaniline, 4-morpholin-4-yl-phenylamine, 4-morpholin-4-yl aniline, n-4-aminophenyl morpholine, p-morpholinoaniline, 4-morpholinobenzenamine, 4-4-morpholino aniline, benzenamine, 4-4-morpholinyl, 4-morpholinyl aniline, 4-4-aminophenyl-morpholine
75655
4-morpholin-4-ylaniline
C1COCCN1C2=CC=C(C=C2)N
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 178.23
Percent Purity 97%
Chemical Name or Material 4-Morpholinoaniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'