missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Morpholineacetic acid hydrochloride, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
4-Morpholineacetic acid hydrochloride is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Morpholineacetic acid hydrochloride is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
4-Morpholineacetic acid hydrochloride is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Color | White |
Quantity | 5 g |
Beilstein | 3701710 |
Solubility Information | Insoluble in water. |
Formula Weight | 181.62 |
Percent Purity | 95% |
Physical Form | Powder |
Chemical Name or Material | 4-Morpholineacetic acid hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'