Få mere at vide
4-Morpholine-d8-carbonyl Chloride, CDN
Artikelnummer. 30319198
Skift visning
Mængde:
0.25 g
Artikelnummer. Mængde
30319198 0.25 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319198 Leverandør CDN Leverandørnr. CDND60520.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Morpholinecarbonyl Chloride-D8
CAS 1219804-24-6
Molekylær formel C5 D8 Cl N O2
Mængde 0.25 g
Synonym 4-Morpholine-d8-carbonyl Chloride, 4-Morpholinecarbonyl Chloride-D8, N-(Morpholine-d8)carbonyl chloride, 4-(Chlorocarbonyl)morpholine-2,2,3,3,5,5,6,6-d8
InChI formel InChI=1 S/C5H8ClNO2/c6-5(8)7-1-3-9-4-2-7/h1-4H2/i1D2,2D2,3D2,4D2
SMIL [2 H]C1([2 H])OC([2 H])([2 H])C([2 H])([2 H])N(C(=O)Cl)C1([2 H])[2 H]
IUPAC navn 2,2,3,3,5,5,6,6-octadeuteriomorpholine-4-carbonyl chloride
Molekylvægt (g/mol) 157.6248
Formel vægt 157.0746 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity