missing translation for 'onlineSavingsMsg'
Få mere at vide

4-morpholin-4-ylbutanoic acid (may contain up to 10% of inorganics), TRC

Artikelnummer. 30342726
Skift visning
Click to view available options
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30342726 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30342726 Supplier Toronto Research Chemicals Supplier No. TRCB4984101G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 4-morpholin-4-ylbutanoic acid
CAS 5807-09-0
Molekylær formel C8 H15 N O3
Mængde 1 g
InChI formel InChI=1S/C8H15NO3/c10-8(11)2-1-3-9-4-6-12-7-5-9/h1-7H2,(H,10,11)
SMIL OC(=O)CCCN1CCOCC1
IUPAC navn 4-morpholin-4-ylbutanoic acid
Molekylvægt (g/mol) 173.21
Formel vægt 173.105
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.